Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Foxk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006247890.2 | XP_006247952.1 | 7 - 8 | 1669 | 136 | |
GenBank mRNA | BC168919.1 | - | 6 - 7 | 1137 | 136 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |