Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aarsd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034109.1 | NP_001029281.1 | 9 - 10 | 962 | 151 | |
GenBank mRNA | BC083790.1 | - | 9 - 10 | 962 | 7106366 | 151 |
FQ213285.1 | - | 9 - 10 | 963 | 151 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |