Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mif4gd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014122.1 | NP_001014144.1 | 3 - 4 | 323 | 62 | |
XM_006247722.1 | XP_006247784.1 | 3 - 4 | 417 | 62 | ||
XM_006247723.2 | XP_006247785.1 | 2 - 3 | 215 | 62 | ||
GenBank mRNA | BC079310.1 | - | 3 - 4 | 323 | 7134761 | 62 |
FQ227422.1 | - | 3 - 4 | 417 | 62 | ||
FQ231060.1 | - | 3 - 4 | 202 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |