Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sept9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113497.1 | NP_001106969.1 | 720 | 101 | ||
NM_031837.2 | NP_114025.2 | 1608 | 101 | |||
NM_176856.2 | NP_789826.2 | 1727 | 101 | |||
GenBank mRNA | AF180525.1 | - | 5 | 887 | 101 | |
AF180526.1 | - | 708 | 101 | |||
BC166602.1 | - | 1273 | 101 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Amplicon spans exons and probe does not span exons |