Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gps2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017477.1 | NP_001017477.1 | 9 - 10 | 1188 | 134 | |
XM_006246785.1 | XP_006246847.1 | 10 - 11 | 1138 | 134 | ||
XM_008767856.1 | XP_008766078.1 | 9 - 10 | 1213 | 134 | ||
GenBank mRNA | BC083699.1 | - | 9 - 10 | 1188 | 7190067 | 134 |
FQ233878.1 | - | 10 - 11 | 1022 | 134 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Inventoried Probe spans exons |