Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clk4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013041.1 | NP_001013059.1 | 9 - 10 | 1072 | 98 | |
XM_006246234.2 | XP_006246296.1 | 10 - 11 | 1282 | 98 | ||
XM_006246236.1 | XP_006246298.1 | 13 - 14 | 1485 | 98 | ||
XM_006246237.1 | XP_006246299.1 | 11 - 12 | 1187 | 98 | ||
XM_006246238.2 | XP_006246300.1 | 10 - 11 | 2702 | 98 | ||
XM_008767670.1 | XP_008765892.1 | 11 - 12 | 2156 | 98 | ||
XM_008767671.1 | XP_008765893.1 | 10 - 11 | 2078 | 98 | ||
XM_008767672.1 | XP_008765894.1 | 10 - 11 | 1121 | 98 | ||
XM_008767673.1 | XP_008765895.1 | 9 - 10 | 2774 | 98 | ||
XM_008767674.1 | XP_008765896.1 | 8 - 9 | 2561 | 98 | ||
XM_008767675.1 | XP_008765897.1 | 11 - 12 | 1348 | 98 | ||
XM_008767676.1 | XP_008765898.1 | 9 - 10 | 1073 | 98 | ||
GenBank mRNA | BC079006.1 | - | 9 - 10 | 1072 | 7113075 | 98 |
FQ223275.1 | - | 12 - 13 | 3132 | 98 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |