Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnf43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001135921.1 | NP_001129393.1 | 2 - 3 | 380 | 125 | |
XM_006247090.2 | XP_006247152.1 | 3 - 4 | 1865 | 125 | ||
XM_006247091.2 | XP_006247153.1 | 3 - 4 | 1640 | 125 | ||
XM_006247092.2 | XP_006247154.1 | 2 - 3 | 2385 | 125 | ||
XM_008768038.1 | XP_008766260.1 | 2 - 3 | 2776 | 125 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |