Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnf43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001135921.1 | NP_001129393.1 | 5 - 6 | 691 | 98 | |
XM_006247090.2 | XP_006247152.1 | 6 - 7 | 2176 | 98 | ||
XM_006247091.2 | XP_006247153.1 | 6 - 7 | 1951 | 98 | ||
XM_006247092.2 | XP_006247154.1 | 5 - 6 | 2696 | 98 | ||
XM_006247094.2 | XP_006247156.1 | 4 - 5 | 2052 | 98 | ||
XM_008768038.1 | XP_008766260.1 | 5 - 6 | 3087 | 98 | ||
XM_008768039.1 | XP_008766261.1 | 3 - 4 | 2730 | 98 | ||
XM_008768040.1 | XP_008766262.1 | 4 - 5 | 2632 | 98 | ||
XM_008768041.1 | XP_008766263.1 | 4 - 5 | 2280 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |