Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Itm2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025712.1 | NP_001020883.1 | 1 - 2 | 217 | 76 | |
GenBank mRNA | BC099174.1 | - | 1 - 2 | 217 | 7382658 | 76 |
FQ216632.1 | - | 1 - 2 | 218 | 76 | ||
FQ217144.1 | - | 1 - 2 | 236 | 76 | ||
FQ224887.1 | - | 1 - 2 | 236 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |