Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Itm2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025712.1 | NP_001020883.1 | 2 - 3 | 347 | 68 | |
GenBank mRNA | BC099174.1 | - | 2 - 3 | 347 | 7382658 | 68 |
FQ216632.1 | - | 2 - 3 | 348 | 68 | ||
FQ217144.1 | - | 2 - 3 | 366 | 68 | ||
FQ224887.1 | - | 2 - 3 | 366 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |