Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Btk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007798.1 | NP_001007799.1 | 6 - 7 | 781 | 93 | |
GenBank mRNA | AY787790.1 | - | 6 - 7 | 781 | 93 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Apoptosis Protein Kinase Detect Genomic DNA Rattus norvegicus Probe spans exons |