Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Khk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031855.3 | NP_114061.1 | 1 - 2 | 293 | 90 | |
XM_006239790.2 | XP_006239852.1 | 1 - 2 | 493 | 90 | ||
XM_008764507.1 | XP_008762729.1 | 1 - 2 | 491 | 90 | ||
GenBank mRNA | BC078752.1 | - | 1 - 2 | 293 | 7109062 | 90 |
X63658.1 | - | 1 - 2 | 353 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |