Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abcg8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_130414.2 | NP_569098.2 | 10 - 11 | 1591 | 91 | |
XM_008764453.1 | XP_008762675.1 | 8 - 9 | 1632 | 91 | ||
GenBank mRNA | AF351785.2 | - | 10 - 11 | 1591 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Transporter Detect Genomic DNA Rattus norvegicus Probe spans exons |