Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trappc6b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106733.1 | NP_001100203.1 | 4 - 5 | 529 | 78 | |
GenBank mRNA | FQ230024.1 | - | 4 - 5 | 292 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Rattus norvegicus Probe spans exons |