Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp6v1c2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014199.1 | NP_001014221.1 | 13 - 14 | 1320 | 65 | |
GenBank mRNA | AY594319.1 | - | 12 - 13 | 1245 | 65 | |
BC079083.1 | - | 13 - 14 | 1320 | 7122992 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Transporter 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |