Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pkdcc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108007.1 | NP_001101477.1 | 6 - 7 | 1007 | 90 | |
GenBank mRNA | BC166535.1 | - | 6 - 7 | 979 | 7191008 | 90 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |