Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp5s | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007749.1 | NP_001007750.1 | 3 - 4 | 629 | 130 | |
XM_006240174.2 | XP_006240236.1 | 3 - 4 | 532 | 130 | ||
XM_006240175.2 | XP_006240237.1 | 3 - 4 | 421 | 130 | ||
XM_006240176.2 | XP_006240238.1 | 3 - 4 | 371 | 130 | ||
GenBank mRNA | BC083655.1 | - | 3 - 4 | 629 | 7135201 | 130 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Transporter Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus Inventoried Probe spans exons |