Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp5s | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007749.1 | NP_001007750.1 | 4 - 5 | 798 | 124 | |
XM_006240174.2 | XP_006240236.1 | 4 - 5 | 701 | 124 | ||
XM_006240175.2 | XP_006240237.1 | 4 - 5 | 590 | 124 | ||
XM_006240176.2 | XP_006240238.1 | 4 - 5 | 540 | 124 | ||
GenBank mRNA | BC083655.1 | - | 4 - 5 | 798 | 7135201 | 124 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Transporter 3' Most Mammalian Gene Collection Rattus norvegicus 1700 Inventoried Probe spans exons |