Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Churc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106741.2 | NP_001100211.1 | 2 - 3 | 230 | 140 | |
GenBank mRNA | BC168770.1 | - | 2 - 3 | 230 | 6918115 | 140 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |