Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Agr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106725.1 | NP_001100195.1 | 2 - 3 | 367 | 124 | |
XM_006240041.2 | XP_006240103.1 | 2 - 3 | 369 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |