Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tsx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019203.1 | NP_062076.1 | 2 - 3 | 198 | 62 | |
XM_006257142.2 | XP_006257204.1 | 3 - 4 | 278 | 62 | ||
GenBank mRNA | X99797.1 | - | 2 - 3 | 198 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons |