Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aldh3b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006998.1 | NP_001006999.1 | 7 - 8 | 991 | 111 | |
XM_006230787.2 | XP_006230849.1 | 8 - 9 | 1294 | 111 | ||
XM_008760132.1 | XP_008758354.1 | 7 - 8 | 1481 | 111 | ||
XM_008760133.1 | XP_008758355.1 | 7 - 8 | 1481 | 111 | ||
XM_008760134.1 | XP_008758356.1 | 7 - 8 | 975 | 111 | ||
GenBank mRNA | BC083850.1 | - | 7 - 8 | 991 | 7122451 | 111 |
FQ212890.1 | - | 1 - 2 | 106 | 111 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |