Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Glyatl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AB019693.1 | - | 93 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus 1700 Probe spans exons |