Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Drd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017140.1 | NP_058836.1 | 3 - 4 | 603 | 114 | |
XM_006248313.2 | XP_006248375.2 | 4 - 5 | 820 | 114 | ||
GenBank mRNA | X53944.1 | - | 3 - 4 | 603 | 114 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor Rattus norvegicus Probe spans exons |