Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adcy5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022600.1 | NP_072122.1 | 11 - 12 | 2787 | 76 | |
XM_008768744.1 | XP_008766966.1 | 11 - 12 | 2960 | 76 | ||
GenBank mRNA | M96159.1 | - | 11 - 12 | 2787 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |