Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mfi2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105872.1 | NP_001099342.1 | 11 - 12 | 1646 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |