Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mfi2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105872.1 | NP_001099342.1 | 12 - 13 | 1803 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons |