Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eaf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172047.1 | NP_742044.1 | 4 - 5 | 617 | 142 | |
XM_008768707.1 | XP_008766929.1 | 4 - 5 | 504 | 142 | ||
XM_008768708.1 | XP_008766930.1 | 2 - 3 | 1173 | 142 | ||
XM_008768709.1 | XP_008766931.1 | 2 - 3 | 903 | 142 | ||
GenBank mRNA | AY049022.1 | - | 4 - 5 | 617 | 142 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |