Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Umps | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025402.1 | NP_001020573.1 | 2 - 3 | 314 | 74 | |
GenBank mRNA | BC098033.1 | - | 2 - 3 | 314 | 7456468 | 74 |
FQ233727.1 | - | 1 - 2 | 52 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |