Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002727893.3 | XP_002727939.1 | 6 - 7 | 717 | 92 | |
XM_006248441.2 | XP_006248503.1 | 2 - 3 | 1231 | 92 | ||
XM_008768770.1 | XP_008766992.1 | 2 - 3 | 1146 | 92 | ||
XM_008768771.1 | XP_008766993.1 | 2 - 3 | 295 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Rattus norvegicus Probe spans exons |