Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gltscr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106226.2 | NP_001099696.2 | 8 - 9 | 3207 | 126 | |
XM_006228343.2 | XP_006228405.1 | 10 - 11 | 3554 | 126 | ||
XM_006228344.2 | XP_006228406.1 | 9 - 10 | 3488 | 126 | ||
XM_006228345.2 | XP_006228407.1 | 9 - 10 | 3377 | 126 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |