Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Uevld | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008759461.1 | XP_008757683.1 | 2 - 3 | 264 | 149 | |
XM_008759462.1 | XP_008757684.1 | 2 - 3 | 263 | 149 | ||
XM_008759463.1 | XP_008757685.1 | 2 - 3 | 259 | 149 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |