Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tyrobp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212525.1 | NP_997690.1 | 3 - 4 | 249 | 72 | |
XM_008759166.1 | XP_008757388.1 | 3 - 4 | 309 | 72 | ||
GenBank mRNA | AY247021.1 | - | 3 - 4 | 249 | 72 | |
FQ211365.1 | - | 3 - 4 | 255 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Rattus norvegicus Probe spans exons |