Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc7a10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053726.2 | NP_446178.2 | 3 - 4 | 578 | 58 | |
XM_006228873.2 | XP_006228935.1 | 3 - 4 | 656 | 58 | ||
XM_006228874.2 | XP_006228936.1 | 3 - 4 | 656 | 58 | ||
GenBank mRNA | AB126813.1 | - | 3 - 4 | 578 | 58 | |
BC127467.1 | - | 3 - 4 | 549 | 7313174 | 58 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |