Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dedd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003748789.2 | XP_003748837.1 | 3 - 4 | 614 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Apoptosis Rattus norvegicus Probe spans exons |