Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc8a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_078619.1 | NP_511174.1 | 2 - 3 | 1338 | 104 | |
GenBank mRNA | U08141.1 | - | 2 - 3 | 1338 | 104 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Transporter Rattus norvegicus Probe spans exons |