Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fxyd5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270688.1 | NP_001257617.1 | 3 - 4 | 170 | 101 | |
NM_001270689.1 | NP_001257618.1 | 3 - 4 | 198 | 101 | ||
NM_021909.2 | NP_068709.2 | 3 - 4 | 274 | 101 | ||
GenBank mRNA | BC060571.1 | - | 3 - 4 | 162 | 6891214 | 101 |
FM112735.1 | - | 3 - 4 | 101 | |||
FM122563.1 | - | 3 - 4 | 148 | 101 | ||
FQ220058.1 | - | 3 - 4 | 198 | 101 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Ion Channel Mammalian Gene Collection Rattus norvegicus Probe spans exons |