Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fuz | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037646.1 | NP_001032735.1 | 3 - 4 | 458 | 81 | |
XM_006229059.2 | XP_006229121.1 | 3 - 4 | 497 | 81 | ||
XM_006229060.2 | XP_006229122.1 | 3 - 4 | 497 | 81 | ||
XM_006229061.2 | XP_006229123.1 | 3 - 4 | 550 | 81 | ||
XM_008759373.1 | XP_008757595.1 | 3 - 4 | 498 | 81 | ||
XM_008759374.1 | XP_008757596.1 | 3 - 4 | 498 | 81 | ||
GenBank mRNA | BC107809.1 | - | 3 - 4 | 458 | 7453736 | 81 |
FQ228777.1 | - | 3 - 4 | 443 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |