Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Numbl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006228565.2 | XP_006228627.1 | 6 - 7 | 3892 | 140 | |
XM_006228567.2 | XP_006228629.1 | 7 - 8 | 951 | 140 | ||
XM_006228568.2 | XP_006228630.1 | 7 - 8 | 1157 | 140 | ||
GenBank mRNA | BC129073.1 | - | 7 - 8 | 838 | 7189989 | 140 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |