Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Npas1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107479.1 | NP_001100949.1 | 1 - 2 | 184 | 69 | |
XM_006228413.2 | XP_006228475.1 | 2 - 3 | 312 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus Probe spans exons |