Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cnfn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108909.1 | NP_001102379.1 | 3 - 4 | 387 | 59 | |
XM_008758960.1 | XP_008757182.1 | 2 - 3 | 433 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Rattus norvegicus Probe spans exons |