Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cnfn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108909.1 | NP_001102379.1 | 4 - 5 | 524 | 101 | |
XM_008758960.1 | XP_008757182.1 | 3 - 4 | 570 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |