Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klk8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107509.1 | NP_001100979.1 | 2 - 3 | 695 | 95 | |
XM_006229049.2 | XP_006229111.1 | 3 - 4 | 296 | 95 | ||
GenBank mRNA | AJ005641.1 | - | 2 - 3 | 363 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Protease Detect Genomic DNA Rattus norvegicus Probe spans exons |