Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klk8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107509.1 | NP_001100979.1 | 3 - 4 | 965 | 105 | |
XM_006229049.2 | XP_006229111.1 | 4 - 5 | 566 | 105 | ||
GenBank mRNA | AJ005641.1 | - | 3 - 4 | 633 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Protease 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |