Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem50b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025014.1 | NP_001020185.1 | 2 - 3 | 257 | 85 | |
XM_006248066.2 | XP_006248128.1 | 3 - 4 | 425 | 85 | ||
XM_006248067.2 | XP_006248129.1 | 3 - 4 | 511 | 85 | ||
XM_006248068.2 | XP_006248130.1 | 2 - 3 | 377 | 85 | ||
XM_006248069.2 | XP_006248131.1 | 2 - 3 | 263 | 85 | ||
GenBank mRNA | BC091349.1 | - | 2 - 3 | 257 | 7316764 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |