Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem50b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025014.1 | NP_001020185.1 | 1 - 2 | 118 | 58 | |
GenBank mRNA | BC091349.1 | - | 1 - 2 | 118 | 7316764 | 58 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons |