Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc5a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053715.2 | NP_446167.2 | 1 | 2069 | 94 | |
GenBank mRNA | AY231162.1 | - | 1 | 2010 | 94 | |
BE116021.1 | - | 1 | 94 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Transporter 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Both primers and probe map within a single exon |