Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrpl39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008768604.1 | XP_008766826.1 | 3 | 465 | 78 | |
GenBank mRNA | FQ225027.1 | - | 3 | 424 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Amplicon spans exons and probe does not span exons |