Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrpl39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008768604.1 | XP_008766826.1 | 7 - 8 | 1015 | 65 | |
GenBank mRNA | FQ225027.1 | - | 7 - 8 | 974 | 65 | |
FQ230552.1 | - | 7 - 8 | 975 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Rattus norvegicus 1700 Probe spans exons |