Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Setd8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001246665.1 | NP_001233594.1 | 4 - 5 | 509 | 77 | |
XM_006249328.2 | XP_006249390.1 | 5 - 6 | 671 | 77 | ||
XM_006249329.2 | XP_006249391.1 | 5 - 6 | 641 | 77 | ||
XM_006249330.2 | XP_006249392.1 | 4 - 5 | 586 | 77 | ||
XM_008769264.1 | XP_008767486.1 | 4 - 5 | 593 | 77 | ||
GenBank mRNA | AB606422.1 | - | 4 - 5 | 509 | 77 | |
DY311222.1 | - | 3 - 4 | 462 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Rattus norvegicus Probe spans exons |